ENA497678264 Enamine

4-oxo-3,4-dihydro-2H-1-benzopyran-2-carboxylic acid

  • MDL number MFCD09262291
  • Cas Number : 51048-00-1
  • Molecular Weight : 192.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-213255

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-213255
Availability details for
Quantity
  • Availability
  • Add to Cart