ENA497678274 Enamine

methyl 2-(propan-2-yloxy)acetate

  • MDL number MFCD10692793
  • Cas Number : 17639-73-5
  • Molecular Weight : 132.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form liquid
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-216374

Safety Information

signalword Warning
pcodes P210,P233,P240,P241,P242,P243,P280,P303+P361+P353,P370+P378,P403+P235,P501
MSDS https://www.enaminestore.com/catalog/EN300-216374
pictograms GHS02
hstatements H226
Availability details for
Quantity
  • Availability
  • Add to Cart