ENA497678395 Enamine

1-(bromomethyl)-1-methanesulfonylcyclohexane

  • MDL number MFCD29762700
  • Cas Number : 1909336-04-4
  • Molecular Weight : 255.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-250590

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-250590
Availability details for
Quantity
  • Availability
  • Add to Cart