ENA497678453 Enamine

isoquinoline-1-thiol

  • MDL number MFCD00160671
  • Cas Number : 4702-25-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 4
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-253645

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-253645
Availability details for
Quantity
  • Availability
  • Add to Cart