ENA497678605 Enamine

2-(1-methylpyrrolidin-3-yl)ethan-1-ol

  • MDL number MFCD17015913
  • Cas Number : 36763-96-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-110052

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-110052
Availability details for
Quantity
  • Availability
  • Add to Cart