ENA497678704 Enamine

5-methoxyisoquinoline-8-carboxylic acid

  • MDL number MFCD22100101
  • Cas Number : 1421600-97-6
  • Molecular Weight : 203.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-258013

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-258013
Availability details for
Quantity
  • Availability
  • Add to Cart