ENA497678741 Enamine

2-(1-aminoethyl)-N,N-dimethylaniline dihydrochloride

  • MDL number MFCD29906870
  • Cas Number : 1909336-50-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data 2HCl
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-258536

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-258536
Availability details for
Quantity
  • Availability
  • Add to Cart