ENA497679261 Enamine

1-phenyl-2-azaspiro[3.4]oct-6-ene

  • MDL number MFCD29763281
  • Cas Number : 1909327-70-3
  • Molecular Weight : 185.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 1-phenyl-2-azaspiro[3.4]oct-6-ene
Inchi Code 1S/C13H15N/c1-2-6-11(7-3-1)12-13(10-14-12)8-4-5-9-13/h1-7,12,14H,8-10H2
Inchi Key CJYDNQIUMXIPAV-UHFFFAOYSA-N

Description

Purity 95
Storage temp. +4
Physical Form liquid
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-262231

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-262231
Availability details for
Quantity
  • Availability
  • Add to Cart