ENA497679270 Enamine

1-(3-methyl-1H-1,2,4-triazol-5-yl)piperazine dihydrochloride

  • MDL number MFCD28895495
  • Molecular Weight : 240.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 1-(3-methyl-1H-1,2,4-triazol-5-yl)piperazine dihydrochloride
Inchi Code 1S/C7H13N5.2ClH/c1-6-9-7(11-10-6)12-4-2-8-3-5-12;;/h8H,2-5H2,1H3,(H,9,10,11);2*1H
Inchi Key PANGMBMOLHRXGW-UHFFFAOYSA-N

Description

Purity 95
Salt Data Cl
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-262317

Safety Information

signalword Warning
pcodes P261,P264,P270,P271,P280,P301+P312,P302+P352,P304+P340,P305+P351+P338,P312,P330,P332+P313,P337+P313,P362,P403+P233,P405,P501
MSDS https://www.enaminestore.com/catalog/EN300-262317
pictograms GHS07
hstatements H302,H315,H319,H335
Availability details for
Quantity
  • Availability
  • Add to Cart