ENA497679271 Enamine

lithium(1+) ion 2-[3-(5-bromothiophen-2-yl)-1,2,4-oxadiazol-5-yl]acetate

  • MDL number MFCD29763291
  • Cas Number : 1909320-04-2
  • Molecular Weight : 295.04

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data Li+
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-262359

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-262359
Availability details for
Quantity
  • Availability
  • Add to Cart