ENA497679278 Enamine

4-[(2-methylthiomorpholin-4-yl)methyl]phenol hydrochloride

  • MDL number MFCD29763301
  • Cas Number : 1909317-53-8
  • Molecular Weight : 259.8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 4-((2-methylthiomorpholino)methyl)phenol hydrochloride
Inchi Code 1S/C12H17NOS.ClH/c1-10-8-13(6-7-15-10)9-11-2-4-12(14)5-3-11;/h2-5,10,14H,6-9H2,1H3;1H
Inchi Key FIRBMRWZMQSXDK-UHFFFAOYSA-N

Description

Purity 95
Salt Data Cl
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-262475

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-262475
Availability details for
Quantity
  • Availability
  • Add to Cart