ENA497679281 Enamine

2-(1-methyl-1H-1,2,3-triazol-4-yl)propan-2-amine dihydrochloride

  • MDL number MFCD29763303
  • Cas Number : 1909309-87-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data 2HCl
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-262503

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-262503
Availability details for
Quantity
  • Availability
  • Add to Cart