ENA497679285 Enamine

(3S)-5-methyl-3-{[(piperidin-2-yl)formamido]methyl}hexanoic acid hydrochloride

  • MDL number MFCD29763306
  • Cas Number : 2103005-40-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. 0
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-262523

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-262523
Availability details for
Quantity
  • Availability
  • Add to Cart