ENA497679288 Enamine

tert-butyl N-[(6S)-2-formyl-4,5,6,7-tetrahydro-1,3-benzothiazol-6-yl]carbamate

  • MDL number MFCD29763310
  • Cas Number : 1909294-18-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-262553

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-262553
Availability details for
Quantity
  • Availability
  • Add to Cart