ENA497679306 Enamine

4-(chloromethyl)-2-(5-methylfuran-2-yl)-1,3-thiazole hydrochloride

  • MDL number MFCD29763331
  • Cas Number : 1909336-77-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. 0
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-262699

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-262699
Availability details for
Quantity
  • Availability
  • Add to Cart