ENA497679311 Enamine

3-bromo-2-(tert-butylsulfanyl)benzaldehyde

  • MDL number MFCD29763335
  • Cas Number : 1326713-90-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-262770

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-262770
Availability details for
Quantity
  • Availability
  • Add to Cart