ENA497679321 Enamine

[(1-methylcyclobutyl)methyl]hydrazine dihydrochloride

  • MDL number MFCD29763345
  • Cas Number : 1909314-34-6
  • Molecular Weight : 187.11

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data 2HCl
Storage temp. +4
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-262829

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-262829
Availability details for
Quantity
  • Availability
  • Add to Cart