ENA497679324 Enamine

1,2,3,5-tetramethylpiperazine

  • MDL number MFCD29907067
  • Cas Number : 1909336-79-3
  • Molecular Weight : 142.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form liquid
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-262911

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-262911
Availability details for
Quantity
  • Availability
  • Add to Cart