ENA497679325 Enamine

2-[(3-chlorophenyl)methyl]-2-methylpyrrolidine hydrochloride

  • MDL number MFCD29763349
  • Cas Number : 1909312-86-2
  • Molecular Weight : 246.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-262934

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-262934
Availability details for
Quantity
  • Availability
  • Add to Cart