ENA497679331 Enamine

(4-methylcyclohexyl)(2-methylphenyl)methanamine hydrochloride

  • MDL number MFCD29763355
  • Cas Number : 1909326-83-5
  • Molecular Weight : 253.81

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-262997

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-262997
Availability details for
Quantity
  • Availability
  • Add to Cart