ENA497679340 Enamine

5-bromo-N-methyl-1,3-thiazol-2-amine hydrobromide

  • MDL number MFCD29763370
  • Cas Number : 1909320-12-2
  • Molecular Weight : 273.98

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HBr
Storage temp. +4
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-263169

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-263169
Availability details for
Quantity
  • Availability
  • Add to Cart