ENA497679344 Enamine

2-{[3-(bromomethyl)phenoxy]methyl}pyridine hydrobromide

  • MDL number MFCD29763375
  • Cas Number : 1909320-11-1
  • Molecular Weight : 359.06

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HBr
Storage temp. +4
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-263264

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-263264
Availability details for
Quantity
  • Availability
  • Add to Cart