ENA497679345 Enamine

2-phenyl-4H,5H,6H,7H-furo[3,2-c]pyridine hydrochloride

  • MDL number MFCD29763376
  • Cas Number : 1909310-10-6
  • Molecular Weight : 235.71

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-263265

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-263265
Availability details for
Quantity
  • Availability
  • Add to Cart