ENA497679580 Enamine

1H-1,2,3,4-tetrazole-5-carbohydrazide

  • MDL number MFCD23884265
  • Cas Number : 1524723-27-0
  • Molecular Weight : 128.09

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-261985

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-261985
Availability details for
Quantity
  • Availability
  • Add to Cart