ENA497679651 Enamine

methyl 5-(3-hydroxyphenyl)-1,2-oxazole-3-carboxylate

  • MDL number MFCD24720352
  • Cas Number : 60640-64-4
  • Molecular Weight : 219.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-262102

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-262102
Availability details for
Quantity
  • Availability
  • Add to Cart