ENA497679695 Enamine

5-chloro-3-(3,5-difluorophenyl)-1,2,4-thiadiazole

  • MDL number MFCD21190941
  • Cas Number : 1491356-45-6
  • Molecular Weight : 232.64

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 5-chloro-3-(3,5-difluorophenyl)-1,2,4-thiadiazole
Inchi Code 1S/C8H3ClF2N2S/c9-8-12-7(13-14-8)4-1-5(10)3-6(11)2-4/h1-3H
Inchi Key REEYZNQDZUQGPF-UHFFFAOYSA-N

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-262264

Safety Information

signalword Warning
pcodes P261,P264,P270,P271,P280,P301+P312,P302+P352,P304+P340,P305+P351+P338,P312,P330,P332+P313,P337+P313,P362,P363,P403+P233,P405,P501
MSDS https://www.enaminestore.com/catalog/EN300-262264
pictograms GHS07
hstatements H302,H312,H315,H319,H332,H335
Availability details for
Quantity
  • Availability
  • Add to Cart