ENA497679696 Enamine

2-amino-6-fluoro-1H-indole-3-carbonitrile

  • MDL number MFCD20227201
  • Molecular Weight : 175.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 2-amino-6-fluoro-1H-indole-3-carbonitrile
Inchi Code 1S/C9H6FN3/c10-5-1-2-6-7(4-11)9(12)13-8(6)3-5/h1-3,13H,12H2
Inchi Key FUZFZIRNBBSCPK-UHFFFAOYSA-N

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-262265

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-262265
Availability details for
Quantity
  • Availability
  • Add to Cart