ENA497679701 Enamine

3-(4-bromophenoxy)aniline

  • MDL number MFCD23979636
  • Cas Number : 141839-57-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-262279

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-262279
Availability details for
Quantity
  • Availability
  • Add to Cart