ENA497679707 Enamine

5-(cyclohexylsulfanyl)-1,2,3-thiadiazole

  • MDL number MFCD00202377
  • Cas Number : 338420-18-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-262285

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-262285
Availability details for
Quantity
  • Availability
  • Add to Cart