ENA497679708 Enamine

3-(oxan-4-yl)propane-1-sulfonyl chloride

  • MDL number MFCD29763287
  • Cas Number : 1504599-35-2
  • Molecular Weight : 226.72

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 3-(tetrahydro-2H-pyran-4-yl)propane-1-sulfonyl chloride
Inchi Code 1S/C8H15ClO3S/c9-13(10,11)7-1-2-8-3-5-12-6-4-8/h8H,1-7H2
Inchi Key PZOPEGDOQPYUSC-UHFFFAOYSA-N

Description

Purity 95
Storage temp. -10
Physical Form oil
Shipping Temperature Ice Pack
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-262287

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-262287
Availability details for
Quantity
  • Availability
  • Add to Cart