ENA497679710 Enamine

3-(2-hydroxyethyl)-1,1-dimethylurea

  • MDL number MFCD21641724
  • Cas Number : 65869-66-1
  • Molecular Weight : 132.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form oil
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-262293

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-262293
Availability details for
Quantity
  • Availability
  • Add to Cart