ENA497679721 Enamine

2-(4-methoxybenzoyl)pyridine

  • MDL number MFCD01716432
  • Cas Number : 6305-18-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-262311

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-262311
Availability details for
Quantity
  • Availability
  • Add to Cart