ENA497679729 Enamine

6,8-dimethyl-3,4-dihydro-2H-1,4-benzoxazin-3-one

  • MDL number MFCD09064401

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-262360

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-262360
Availability details for
Quantity
  • Availability
  • Add to Cart