ENA497679731 Enamine

3-[1-hydroxy-2-(methylamino)ethyl]benzonitrile

  • MDL number MFCD20545904
  • Cas Number : 672326-25-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-262371

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-262371
Availability details for
Quantity
  • Availability
  • Add to Cart