ENA497679732 Enamine

4-methyl-5-(phenylsulfanyl)-1,2,3-thiadiazole

  • MDL number MFCD00202247
  • Cas Number : 147123-18-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-262374

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-262374
Availability details for
Quantity
  • Availability
  • Add to Cart