ENA497679740 Enamine

4-ethynyl-2,3-dihydro-1H-isoindole-1,3-dione

  • MDL number MFCD29763294
  • Cas Number : 1909309-81-4
  • Molecular Weight : 171.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-262391

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-262391
Availability details for
Quantity
  • Availability
  • Add to Cart