ENA497679746 Enamine

5-bromo-4-chloro-2-methoxypyrimidine

  • MDL number MFCD12400758
  • Cas Number : 57054-93-0
  • Molecular Weight : 223.46

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 5-bromo-4-chloro-2-methoxypyrimidine
Inchi Code 1S/C5H4BrClN2O/c1-10-5-8-2-3(6)4(7)9-5/h2H,1H3
Inchi Key CUJKVRNKYMBUGX-UHFFFAOYSA-N

Description

Purity 95
Storage temp. +4
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-262400

Safety Information

signalword Warning
pcodes P261,P264,P270,P271,P280,P301+P312,P302+P352,P304+P340,P305+P351+P338,P312,P330,P332+P313,P337+P313,P362,P403+P233,P405,P501
MSDS https://www.enaminestore.com/catalog/EN300-262400
pictograms GHS07
hstatements H302,H315,H319,H335
Availability details for
Quantity
  • Availability
  • Add to Cart