ENA497679748 Enamine

5-chloro-2-(trifluoromethoxy)benzene-1-sulfonamide

  • MDL number MFCD18393787
  • Cas Number : 77797-66-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-262402

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-262402
Availability details for
Quantity
  • Availability
  • Add to Cart