ENA497679759 Enamine

4-amino-1-methyl-N-(1,2-oxazol-4-yl)-1H-pyrazole-3-carboxamide

  • MDL number MFCD21221520
  • Cas Number : 1472690-97-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-262424

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-262424
Availability details for
Quantity
  • Availability
  • Add to Cart