ENA497679775 Enamine

4-[4-(2,4,4-trimethylpentan-2-yl)phenoxy]butan-1-ol

  • MDL number MFCD13717388
  • Cas Number : 1153244-40-6
  • Molecular Weight : 278.44

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 4-(4-(2,4,4-trimethylpentan-2-yl)phenoxy)butan-1-ol
Inchi Code 1S/C18H30O2/c1-17(2,3)14-18(4,5)15-8-10-16(11-9-15)20-13-7-6-12-19/h8-11,19H,6-7,12-14H2,1-5H3
Inchi Key UGFYLYJDGQTZSR-UHFFFAOYSA-N

Description

Purity 95
Storage temp. RT
Physical Form oil
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-262459

Safety Information

signalword Warning
pcodes P261,P264,P270,P271,P280,P301+P312,P302+P352,P304+P340,P305+P351+P338,P312,P330,P332+P313,P337+P313,P362,P403+P233,P405,P501
MSDS https://www.enaminestore.com/catalog/EN300-262459
pictograms GHS07
hstatements H302,H315,H319,H335
Availability details for
Quantity
  • Availability
  • Add to Cart