ENA497679800 Enamine

1-(4-methylphenyl)cyclobutan-1-amine hydrochloride

  • MDL number MFCD09864763
  • Cas Number : 1228878-94-1
  • Molecular Weight : 197.71

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 1-(4-methylphenyl)cyclobutanamine hydrochloride
Inchi Code 1S/C11H15N.ClH/c1-9-3-5-10(6-4-9)11(12)7-2-8-11;/h3-6H,2,7-8,12H2,1H3;1H
Inchi Key CVZCXKNLYKJZMI-UHFFFAOYSA-N

Description

Purity 95
Salt Data Cl
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-262541

Safety Information

signalword Warning
pcodes P261,P264,P270,P271,P280,P301+P312,P302+P352,P304+P340,P305+P351+P338,P312,P330,P332+P313,P337+P313,P362,P403+P233,P405,P501
MSDS https://www.enaminestore.com/catalog/EN300-262541
pictograms GHS07
hstatements H302,H315,H319,H335
Availability details for
Quantity
  • Availability
  • Add to Cart