ENA497679809 Enamine

8-thiabicyclo[3.2.1]octan-3-one

  • MDL number MFCD24688193
  • Cas Number : 16892-50-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-262557

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-262557
Availability details for
Quantity
  • Availability
  • Add to Cart