ENA497679811 Enamine

4-bromo-1-chloro-7-fluoroisoquinoline

  • MDL number MFCD11870524
  • Cas Number : 1369199-73-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-262562

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-262562
Availability details for
Quantity
  • Availability
  • Add to Cart