ENA497679815 Enamine

1-tert-butyl-4-iodo-1H-pyrazol-5-amine

  • MDL number MFCD23881340
  • Cas Number : 1499849-59-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-262567

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-262567
Availability details for
Quantity
  • Availability
  • Add to Cart