ENA497679818 Enamine

2-(2-chloro-6-fluorophenyl)morpholine

  • MDL number MFCD09901460
  • Cas Number : 1017480-74-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-262573

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-262573
Availability details for
Quantity
  • Availability
  • Add to Cart