ENA497679822 Enamine

3-cyclopropylprop-2-enal

  • Cas Number : 5687-76-3
  • Molecular Weight : 96.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name (2E)-3-cyclopropyl-2-propenal
Inchi Code 1S/C6H8O/c7-5-1-2-6-3-4-6/h1-2,5-6H,3-4H2/b2-1+
Inchi Key KZZDSVLUZALMOT-OWOJBTEDSA-N

Description

Purity 95
Storage temp. -10
Physical Form liquid
Shipping Temperature Ice Pack
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-262578

Safety Information

signalword Warning
pcodes P210,P261,P264,P271,P280,P302+P352,P304+P340,P305+P351+P338,P312,P332+P313,P337+P313,P362,P370+P378,P403+P233,P403+P235,P405,P501
MSDS https://www.enaminestore.com/catalog/EN300-262578
pictograms GHS07
hstatements H227,H315,H319,H335
Availability details for
Quantity
  • Availability
  • Add to Cart