ENA497679826 Enamine

ethyl 6-methoxy-3-oxohexanoate

  • MDL number MFCD21125644
  • Cas Number : 1227787-88-3
  • Molecular Weight : 188.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name ethyl 6-methoxy-3-oxohexanoate
Inchi Code 1S/C9H16O4/c1-3-13-9(11)7-8(10)5-4-6-12-2/h3-7H2,1-2H3
Inchi Key VUCFQBNPRHCETH-UHFFFAOYSA-N

Description

Purity 95
Storage temp. -10
Physical Form liquid
Shipping Temperature Ice Pack
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-262582

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-262582
Availability details for
Quantity
  • Availability
  • Add to Cart