ENA497679847 Enamine

tert-butyl 2-amino-2-(1-methyl-1H-pyrazol-5-yl)acetate hydrochloride

  • MDL number MFCD29763322
  • Cas Number : 1909309-39-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-262611

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-262611
Availability details for
Quantity
  • Availability
  • Add to Cart