ENA497679852 Enamine

ethyl 2-{[2-(tert-butoxy)-2-oxoethyl]sulfanyl}-2-fluoroacetate

  • MDL number MFCD29907060
  • Cas Number : 1592888-07-7
  • Molecular Weight : 252.31

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name ethyl 2-((2-(tert-butoxy)-2-oxoethyl)thio)-2-fluoroacetate
Inchi Code 1S/C10H17FO4S/c1-5-14-9(13)8(11)16-6-7(12)15-10(2,3)4/h8H,5-6H2,1-4H3
Inchi Key FOVXEGQNMQZSBO-UHFFFAOYSA-N

Description

Purity 95
Storage temp. +4
Physical Form liquid
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-262638

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-262638
Availability details for
Quantity
  • Availability
  • Add to Cart