ENA497679866 Enamine

5-(chloromethyl)-1,2,3-thiadiazole

  • MDL number MFCD16497116
  • Cas Number : 1284385-87-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-262661

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-262661
Availability details for
Quantity
  • Availability
  • Add to Cart